Assay Details
Gene Symbol: NCF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030709.1 | NP_001025880.1 | 4 - 5 | 436 | 94 | |
XM_015295704.2 | XP_015151190.1 | 3 - 4 | 362 | 94 | ||
GenBank mRNA | AJ719555.1 | - | 4 - 5 | 436 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
