Assay Details
| Gene Symbol: NCF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030709.1 | NP_001025880.1 | 9 - 10 | 955 | 68 | |
| XM_015295704.2 | XP_015151190.1 | 8 - 9 | 881 | 68 | ||
| GenBank mRNA | AJ719555.1 | - | 9 - 10 | 955 | 68 | |
More Information
Set Membership: |
|

NCF1C
Genomic Map