Assay Details
| Gene Symbol: DPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030716.2 | NP_001025887.2 | 5 - 6 | 561 | 58 | |
| XM_015295755.2 | XP_015151241.1 | 5 - 6 | 662 | 58 | ||
| GenBank mRNA | AJ721005.1 | - | 5 - 6 | 561 | 58 | |
More Information
Set Membership: |
|

DPH1
Genomic Map