Assay Details
Gene Symbol: TRABD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030732.1 | NP_001025903.1 | 4 - 5 | 442 | 64 | |
XM_015274537.2 | XP_015130023.1 | 4 - 5 | 507 | 64 | ||
XM_015274569.2 | XP_015130055.1 | 4 - 5 | 439 | 64 | ||
GenBank mRNA | AJ851497.1 | - | 4 - 5 | 442 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
