Assay Details
Gene Symbol: IRAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030738.1 | NP_001025909.1 | 4 - 5 | 797 | 66 | |
XM_015281244.2 | XP_015136730.1 | 4 - 5 | 66 | |||
GenBank mRNA | AJ720408.1 | - | 4 - 5 | 797 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
