Assay Details
| Gene Symbol: TMEM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030759.1 | NP_001025930.1 | 2 - 3 | 390 | 61 | |
| XM_015285263.2 | XP_015140749.1 | 2 - 3 | 419 | 61 | ||
| XM_015285273.2 | XP_015140759.1 | 2 - 3 | 426 | 61 | ||
| XM_015285276.2 | XP_015140762.1 | 2 - 3 | 437 | 61 | ||
| GenBank mRNA | AJ851700.1 | - | 2 - 3 | 390 | 61 | |
More Information
Set Membership: |
|

TMEM184B
Genomic Map