Assay Details
| Gene Symbol: CECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030767.2 | NP_001025938.1 | 3 - 4 | 653 | 58 | |
| XM_015285830.2 | XP_015141316.1 | 3 - 4 | 1210 | 58 | ||
| XM_015285839.2 | XP_015141325.1 | 3 - 4 | 667 | 58 | ||
| GenBank mRNA | AJ720326.1 | - | 3 - 4 | 654 | 58 | |
| AY902779.1 | - | 3 - 4 | 656 | 58 | ||
More Information
Set Membership: |
|

CECR1
Genomic Map