Assay Details
| Gene Symbol: CECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030767.2 | NP_001025938.1 | 8 - 9 | 1355 | 109 | |
| XM_015285830.2 | XP_015141316.1 | 8 - 9 | 1912 | 109 | ||
| XM_015285839.2 | XP_015141325.1 | 8 - 9 | 1369 | 109 | ||
| GenBank mRNA | AJ720326.1 | - | 8 - 9 | 1356 | 109 | |
| AY902779.1 | - | 8 - 9 | 1358 | 109 | ||
More Information
Set Membership: |
|

CECR1
Genomic Map