Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030791.1 | NP_001025962.1 | 4 - 5 | 354 | 78 | |
| XM_025149429.1 | XP_025005197.1 | 4 - 5 | 557 | 78 | ||
| XM_025149433.1 | XP_025005201.1 | 4 - 5 | 638 | 78 | ||
| GenBank mRNA | AJ851452.1 | - | 4 - 5 | 354 | 78 | |
More Information
Set Membership: |
|

GPR89A
Genomic Map