Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030791.1 | NP_001025962.1 | 8 - 9 | 770 | 106 | |
| GenBank mRNA | AJ851452.1 | - | 8 - 9 | 770 | 106 | |
| CR523472.1 | - | 1 - 2 | 120 | 106 | ||
More Information
Set Membership: |
|

GPR89A
Genomic Map