Assay Details
| Gene Symbol: TCEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030905.1 | NP_001026076.1 | 2 - 3 | 135 | 61 | |
| XM_015297671.2 | XP_015153157.1 | 2 - 3 | 227 | 61 | ||
| GenBank mRNA | AJ720846.1 | - | 2 - 3 | 135 | 61 | |
More Information
Set Membership: |
|

TCEB3
Genomic Map