Assay Details
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030911.4 | NP_001026082.2 | 3 - 4 | 269 | 68 | |
XM_015297997.2 | XP_015153483.1 | 3 - 4 | 247 | 68 | ||
XM_025143186.1 | XP_024998954.1 | 3 - 4 | 193 | 68 | ||
GenBank mRNA | AJ448330.1 | - | 3 - 4 | 270 | 68 | |
AJ719312.1 | - | 3 - 4 | 248 | 68 | ||
CN232272.1 | - | 3 - 4 | 262 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
