Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030969.1 | NP_001026140.1 | 3 - 4 | 1152 | 64 | |
| GenBank mRNA | AJ720905.1 | - | 3 - 4 | 1152 | 64 | |
| KY747396.1 | - | 3 - 4 | 830 | 64 | ||
More Information
Set Membership: |
|

KLF6
Genomic Map