Assay Details
| Gene Symbol: PIP4K2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030971.1 | NP_001026142.1 | 9 - 10 | 1308 | 69 | |
| XM_025147179.1 | XP_025002947.1 | 9 - 10 | 1016 | 69 | ||
| XM_025147180.1 | XP_025002948.1 | 7 - 8 | 955 | 69 | ||
| XM_025147181.1 | XP_025002949.1 | 9 - 10 | 1112 | 69 | ||
| GenBank mRNA | AJ851794.1 | - | 9 - 10 | 1308 | 69 | |
More Information
Set Membership: |
|

PIP4K2A
Genomic Map