Assay Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030974.1 | NP_001026145.1 | 3 - 4 | 488 | 81 | |
| GenBank mRNA | AJ851572.1 | - | 3 - 4 | 488 | 81 | |
More Information
Set Membership: |
|

RBM48
Genomic Map