Assay Details
Gene Symbol: PI4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031157.1 | NP_001026328.1 | 9 - 10 | 1370 | 79 | |
XM_015285691.2 | XP_015141177.1 | 9 - 10 | 1359 | 79 | ||
XM_015285692.2 | XP_015141178.1 | 9 - 10 | 1283 | 79 | ||
XM_025149695.1 | XP_025005463.1 | 9 - 10 | 1244 | 79 | ||
XM_025149696.1 | XP_025005464.1 | 9 - 10 | 1819 | 79 | ||
GenBank mRNA | AJ720784.1 | - | 9 - 10 | 1370 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
