Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031166.2 | NP_001026337.2 | 2 - 3 | 804 | 125 | |
| GenBank mRNA | AJ851742.1 | - | 2 - 3 | 810 | 125 | |
More Information
Set Membership: |
|

TNIP2
Genomic Map