Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031166.2 | NP_001026337.2 | 5 - 6 | 1318 | 78 | |
| XM_015285792.2 | XP_015141278.1 | 5 - 6 | 3893 | 78 | ||
| GenBank mRNA | AJ851742.1 | - | 5 - 6 | 1324 | 78 | |
More Information
Set Membership: |
|

TNIP2
Genomic Map