Assay Details
| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031289.1 | NP_001026460.1 | 1 | 1147 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

JUN| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031289.1 | NP_001026460.1 | 1 | 1147 | 74 | |
Set Membership: |
|