Assay Details
| Gene Symbol: ZRANB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031297.1 | NP_001026468.1 | 3 - 4 | 274 | 65 | |
| XM_015291056.2 | XP_015146542.1 | 3 - 4 | 286 | 65 | ||
| GenBank mRNA | AJ719609.1 | - | 3 - 4 | 274 | 65 | |
More Information
Set Membership: |
|

ZRANB2
Genomic Map