Assay Details
| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031343.1 | NP_001026514.1 | 3 - 4 | 502 | 64 | |
| XM_025147313.1 | XP_025003081.1 | 3 - 4 | 730 | 64 | ||
| GenBank mRNA | AJ720155.1 | - | 3 - 4 | 502 | 64 | |
More Information
Set Membership: |
|

YWHAZ
Genomic Map