Assay Details
Gene Symbol: SRSF7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031368.1 | NP_001026539.1 | 4 - 5 | 537 | 108 | |
XM_015283521.2 | XP_015139007.1 | 4 - 5 | 537 | 108 | ||
GenBank mRNA | AJ719404.1 | - | 4 - 5 | 537 | 108 | |
CR385735.1 | - | 1 - 2 | 413 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
