Assay Details
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031375.3 | NP_001026546.2 | 1 - 2 | 335 | 79 | |
XM_015285444.2 | XP_015140930.1 | 1 - 2 | 258 | 79 | ||
XM_015285446.2 | XP_015140932.1 | 1 - 2 | 258 | 79 | ||
GenBank mRNA | AJ720001.1 | - | 1 - 2 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
