Assay Details
Gene Symbol: PSMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031407.1 | NP_001026578.1 | 672 | 66 | ||
GenBank mRNA | AJ720305.1 | - | 672 | 66 | ||
CR386956.1 | - | 683 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
