Assay Details
| Gene Symbol: TXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031410.1 | NP_001026581.1 | 3 - 4 | 368 | 65 | |
| XM_015285949.2 | XP_015141435.1 | 3 - 4 | 376 | 65 | ||
| XM_025151067.1 | XP_025006835.1 | 3 - 4 | 2375 | 65 | ||
| GenBank mRNA | AJ720298.1 | - | 3 - 4 | 368 | 65 | |
| CR385786.1 | - | 436 | 65 | |||
| CR387816.1 | - | 2 - 3 | 339 | 65 | ||
| CR389326.1 | - | 610 | 65 | |||
| CR390224.1 | - | 2 - 3 | 350 | 65 | ||
More Information
Set Membership: |
|

TXN2
Genomic Map