Assay Details
| Gene Symbol: VDAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033869.2 | NP_001029041.1 | 5 - 6 | 433 | 82 | |
| XM_015294017.2 | XP_015149503.1 | 5 - 6 | 511 | 82 | ||
| XM_025154858.1 | XP_025010626.1 | 5 - 6 | 354 | 82 | ||
| GenBank mRNA | CR390115.1 | - | 5 - 6 | 434 | 82 | |
More Information
Set Membership: |
|

VDAC1
Genomic Map