Assay Details
| Gene Symbol: CAMK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034813.1 | NP_001029985.1 | 6 - 7 | 507 | 67 | |
| GenBank mRNA | BX932197.1 | - | 6 - 7 | 569 | 67 | |
| BX934362.2 | - | 6 - 7 | 565 | 67 | ||
| BX950684.2 | - | 6 - 7 | 587 | 67 | ||
More Information
Set Membership: |
|

CAMK4
Genomic Map