Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034825.2 | NP_001029997.2 | 3 - 4 | 502 | 77 | |
| GenBank mRNA | AB193124.1 | - | 3 - 4 | 502 | 77 | |
More Information
Set Membership: |
|

NPFFR2
Genomic Map