Assay Details
| Gene Symbol: NPY7R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037824.1 | NP_001032913.1 | 1 | 1052 | 110 | |
| XM_015293791.2 | XP_015149277.1 | 2 | 1358 | 110 | ||
More Information
Set Membership: |
|

NPY7R
Genomic Map