Assay Details
Gene Symbol: FBXW11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039262.1 | NP_001034351.1 | 4 - 5 | 579 | 72 | |
XM_015293592.2 | XP_015149078.1 | 5 - 6 | 665 | 72 | ||
XM_015293593.2 | XP_015149079.1 | 7 - 8 | 885 | 72 | ||
XM_015293594.2 | XP_015149080.1 | 6 - 7 | 776 | 72 | ||
XM_015293595.2 | XP_015149081.1 | 4 - 5 | 512 | 72 | ||
XM_015293596.2 | XP_015149082.1 | 5 - 6 | 674 | 72 | ||
XM_015293597.2 | XP_015149083.1 | 4 - 5 | 816 | 72 | ||
XM_025154836.1 | XP_025010604.1 | 4 - 5 | 8922 | 72 | ||
XM_025154837.1 | XP_025010605.1 | 5 - 6 | 7813 | 72 | ||
XM_025154838.1 | XP_025010606.1 | 5 - 6 | 8803 | 72 | ||
XM_025154839.1 | XP_025010607.1 | 5 - 6 | 8798 | 72 | ||
GenBank mRNA | AJ721075.1 | - | 4 - 5 | 579 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
