Assay Details
Gene Symbol: COLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039598.1 | NP_001034687.1 | 3 - 4 | 328 | 73 | |
XM_015283021.2 | XP_015138507.1 | 3 - 4 | 428 | 73 | ||
GenBank mRNA | DQ129668.1 | - | 3 - 4 | 328 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
