Assay Details
| Gene Symbol: IFI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001296.5 | NP_001001296.1 | 1 - 2 | 157 | 66 | |
| GenBank mRNA | BX272937.4 | - | 1 - 2 | 157 | 66 | |
More Information
Set Membership: |
|

IFI6
Genomic Map