Assay Details
| Gene Symbol: KRT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001311.2 | NP_001001311.2 | 7 - 8 | 1343 | 79 | |
| GenBank mRNA | AY574987.1 | - | 7 - 8 | 1343 | 79 | |
More Information
Set Membership: |
|

KRT14
Genomic Map