Assay Details
| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006239.3 | NP_001006239.2 | 2 - 3 | 228 | 103 | |
| GenBank mRNA | AJ719419.1 | - | 2 - 3 | 196 | 103 | |
| CR353921.1 | - | 2 - 3 | 228 | 103 | ||
More Information
Set Membership: |
|

RPS19BP1
Genomic Map