Assay Details
| Gene Symbol: N4BP2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277885.2 | XP_015133371.1 | 4 - 5 | 1958 | 65 | |
| XM_015277889.2 | XP_015133375.1 | 4 - 5 | 1940 | 65 | ||
| XM_015277896.2 | XP_015133382.1 | 4 - 5 | 1961 | 65 | ||
| XM_015277900.2 | XP_015133386.1 | 4 - 5 | 1958 | 65 | ||
| XM_015277908.2 | XP_015133394.1 | 4 - 5 | 1956 | 65 | ||
| XM_015277912.2 | XP_015133398.1 | 4 - 5 | 1956 | 65 | ||
| XM_025146830.1 | XP_025002598.1 | 4 - 5 | 1955 | 65 | ||
| GenBank mRNA | AJ721041.1 | - | 4 - 5 | 1712 | 65 | |
| CR390564.1 | - | 3 - 4 | 344 | 65 | ||
More Information
Set Membership: |
|

N4BP2L2
Genomic Map