Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024734.3 | NP_001019905.1 | 4 - 5 | 452 | 140 | |
| GenBank mRNA | AL588177.1 | - | 4 - 5 | 453 | 140 | |
| BX931291.2 | - | 4 - 5 | 415 | 140 | ||
| BX934085.2 | - | 4 - 5 | 440 | 140 | ||
More Information
Set Membership: |
|

SELENOS
Genomic Map