Assay Details
| Gene Symbol: MRPL48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030053.2 | NP_001025224.1 | 5 - 6 | 412 | 69 | |
| XM_015280972.2 | XP_015136458.1 | 5 - 6 | 636 | 69 | ||
| XM_025150714.1 | XP_025006482.1 | 5 - 6 | 399 | 69 | ||
| GenBank mRNA | BU258502.1 | - | 5 - 6 | 386 | 69 | |
| BX934053.2 | - | 5 - 6 | 407 | 69 | ||
| BX934907.1 | - | 5 - 6 | 404 | 69 | ||
| BX950666.1 | - | 5 - 6 | 412 | 69 | ||
More Information
Set Membership: |
|

MRPL48
Genomic Map