Assay Details
| Gene Symbol: IGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030342.3 | NP_001025513.1 | 1 - 2 | 157 | 79 | |
| XM_015286525.2 | XP_015142011.1 | 2 - 3 | 1914 | 79 | ||
| GenBank mRNA | AY267181.1 | - | 1 - 2 | 127 | 79 | |
More Information
Set Membership: |
|

IGF2
Genomic Map