Assay Details
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032390.2 | NP_001027562.2 | 1 - 2 | 67 | 87 | |
GenBank mRNA | BX931608.2 | - | 1 - 2 | 51 | 87 | |
BX933628.2 | - | 1 - 2 | 44 | 87 | ||
BX933641.1 | - | 1 - 2 | 42 | 87 | ||
BX933648.1 | - | 1 - 2 | 49 | 87 | ||
BX933649.2 | - | 1 - 2 | 49 | 87 | ||
BX933653.2 | - | 1 - 2 | 64 | 87 | ||
BX933662.1 | - | 1 - 2 | 32 | 87 | ||
BX933671.1 | - | 1 - 2 | 42 | 87 | ||
BX933686.1 | - | 1 - 2 | 67 | 87 | ||
BX933689.1 | - | 1 - 2 | 61 | 87 | ||
BX935221.1 | - | 1 - 2 | 37 | 87 | ||
BX935254.1 | - | 1 - 2 | 77 | 87 | ||
BX935323.2 | - | 1 - 2 | 49 | 87 | ||
BX936261.2 | - | 1 - 2 | 68 | 87 | ||
CR338993.2 | - | 1 - 2 | 49 | 87 | ||
CR338996.1 | - | 1 - 2 | 51 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
