Assay Details
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032390.2 | NP_001027562.2 | 4 - 5 | 383 | 119 | |
GenBank mRNA | BX933628.2 | - | 3 - 4 | 261 | 119 | |
BX933648.1 | - | 4 - 5 | 365 | 119 | ||
BX933649.2 | - | 4 - 5 | 365 | 119 | ||
BX933653.2 | - | 4 - 5 | 380 | 119 | ||
BX933662.1 | - | 4 - 5 | 348 | 119 | ||
BX933680.2 | - | 4 - 5 | 357 | 119 | ||
BX933686.1 | - | 4 - 5 | 383 | 119 | ||
BX935221.1 | - | 4 - 5 | 353 | 119 | ||
BX935323.2 | - | 4 - 5 | 365 | 119 | ||
BX935324.2 | - | 4 - 5 | 357 | 119 | ||
BX935330.2 | - | 4 - 5 | 345 | 119 | ||
CR338993.2 | - | 4 - 5 | 365 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
