Assay Details
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032390.2 | NP_001027562.2 | 7 - 8 | 817 | 115 | |
XM_025142533.1 | XP_024998301.1 | 3 - 4 | 649 | 115 | ||
GenBank mRNA | BU487365.1 | - | 4 - 5 | 508 | 115 | |
BX933628.2 | - | 6 - 7 | 695 | 115 | ||
BX933648.1 | - | 7 - 8 | 799 | 115 | ||
BX933649.2 | - | 7 - 8 | 799 | 115 | ||
BX933653.2 | - | 7 - 8 | 814 | 115 | ||
BX933662.1 | - | 7 - 8 | 782 | 115 | ||
BX933680.2 | - | 7 - 8 | 791 | 115 | ||
BX933686.1 | - | 7 - 8 | 817 | 115 | ||
BX935221.1 | - | 7 - 8 | 787 | 115 | ||
BX935323.2 | - | 7 - 8 | 799 | 115 | ||
BX935324.2 | - | 7 - 8 | 791 | 115 | ||
BX935330.2 | - | 7 - 8 | 779 | 115 | ||
CR338993.2 | - | 7 - 8 | 799 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025142533.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
