Assay Details
Gene Symbol: LOC395100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039596.1 | NP_001034685.1 | 417 | 82 | ||
XM_015298681.2 | XP_015154167.1 | 566 | 82 | |||
XM_015298682.2 | XP_015154168.1 | 1586 | 82 | |||
GenBank mRNA | AY300013.1 | - | 417 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
