Assay Details
| Gene Symbol: IL10RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039597.1 | NP_001034686.1 | 1 - 2 | 95 | 57 | |
| GenBank mRNA | AM049243.1 | - | 1 - 2 | 95 | 57 | |
| AM049244.1 | - | 1 - 2 | 95 | 57 | ||
More Information
Set Membership: |
|

IL10RA
Genomic Map