Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040091.1 | NP_001035180.1 | 3 - 4 | 647 | 66 | |
| GenBank mRNA | AM236874.1 | - | 3 - 4 | 647 | 66 | |
More Information
Set Membership: |
|

CTLA4
Genomic Map