Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040120.2 | NP_001035209.1 | 2 - 3 | 316 | 129 | |
| GenBank mRNA | AB106886.1 | - | 2 - 3 | 317 | 129 | |
| BX932476.2 | - | 2 - 3 | 198 | 129 | ||
More Information
Set Membership: |
|

BRI3BP
Genomic Map