Assay Details
| Gene Symbol: MAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044671.1 | NP_001038136.1 | 2 - 3 | 1714 | 65 | |
| GenBank mRNA | D28598.1 | - | 2 - 3 | 1714 | 65 | |
More Information
Set Membership: |
|

MAF
Genomic Map