Assay Details
| Gene Symbol: PCDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045827.1 | NP_001039292.1 | 2 - 3 | 882 | 97 | |
| XM_015293599.2 | XP_015149085.1 | 2 - 3 | 97 | |||
| XM_015293600.2 | XP_015149086.1 | 2 - 3 | 97 | |||
| GenBank mRNA | AY676328.1 | - | 2 - 3 | 882 | 97 | |
More Information
Set Membership: |
|

PCDH1
Genomic Map