Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045828.1 | NP_001039293.1 | 4 - 5 | 825 | 58 | |
| XM_025154862.1 | XP_025010630.1 | 4 - 5 | 3541 | 58 | ||
| XM_025154863.1 | XP_025010631.1 | 4 - 5 | 3541 | 58 | ||
| XM_025154864.1 | XP_025010632.1 | 4 - 5 | 3541 | 58 | ||
More Information
Set Membership: |
|

SLC22A5
Genomic Map