Assay Details
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045830.1 | NP_001039295.1 | 2 - 3 | 210 | 94 | |
XM_015279442.2 | XP_015134928.1 | 2 - 3 | 241 | 94 | ||
XM_015279443.2 | XP_015134929.1 | 2 - 3 | 232 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
