Assay Details
| Gene Symbol: IL13RA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048078.1 | NP_001041543.1 | 5 - 6 | 799 | 75 | |
| XM_015278228.2 | XP_015133714.1 | 6 - 7 | 982 | 75 | ||
| XM_015278229.2 | XP_015133715.1 | 6 - 7 | 1230 | 75 | ||
| XM_015278230.2 | XP_015133716.1 | 6 - 7 | 1036 | 75 | ||
| GenBank mRNA | AB243738.1 | - | 5 - 6 | 799 | 75 | |
More Information
Set Membership: |
|

IL13RA2
Genomic Map